missing translation for 'onlineSavingsMsg'
Learn More

(6,7-Dimethoxy-4-Oxoquinazolin-3(4h)-Yl)acetic Acid, TRC

Product Code. 30374019
Change view
Click to view available options
Quantity:
250 mg
500 mg
1 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
30374019 1 g
30379745 250 mg
30372356 500 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30374019 Supplier Toronto Research Chemicals Supplier No. TRCD8309431G

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 877140-09-5
Molecular Formula C12H12N2O5
Molecular Weight (g/mol) 264.23
Synonym 6,7-Dimethoxy-4-oxo-3(4H)-quinazolineacetic Acid, (6,7-Dimethoxy-4-oxoquinazolin-3(4H)-yl)acetic Acid, 2-(6,7-Dimethoxy-4-oxo-3,4-dihydroquinazolin-3-yl)acetic Acid, 2-(6,7-Dimethoxy-4-oxoquinazolin-3-yl)acetic Acid
SMILES COc1cc2N=CN(CC(=O)O)C(=O)c2cc1OC

Specifications

Quantity 1 g
InChI Formula InChI=1S/C12H12N2O5/c1-18-9-3-7-8(4-10(9)19-2)13-6-14(12(7)17)5-11(15)16/h3-4,6H,5H2,1-2H3,(H,15,16)
Formula Weight 264.0746
Chemical Name or Material (6,7-Dimethoxy-4-Oxoquinazolin-3(4h)-Yl)acetic Acid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.