missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Titanium, Ti-6-4-2, R54620, 65g, ARMI∣MBH
Click to view available options
Quantity:
65 g
Unit Size:
65g
Description
Titanium, Ti-6-4-2, R54620, 65g
Specifications
Specifications
| Chemical Name or Material | Hydrogen/Nickel/Nitrogen/Carbon/Iron/Silicon/Molybdenum/Tin/Zirconium/Aluminium/Antimony/Silver/Tantalum/Arsenic/Calcium/Cobalt/Hafnium/Manganese/Tungsten/Yttrium/Boron/Lead/Magnesium/Niobium/Zinc/Vanadium/Chromium/Copper/Phosphorus/Oxygen/Titanium |
| CAS | 7440-32-6 |
| Quantity | 65 g |
| Form | Pieces |
| Concentration or Composition (by Analyte or Components) | Hydrogen 0.0022 %/Nickel 0.0038 %/Nitrogen 0.004 %/Carbon 0.007 %/Iron 0.025 %/Silicon 0.077 %/Molybdenum 2.02 %/Tin 2.06 %/Zirconium 4.05 %/Aluminium 6.11 %/Antimony (<0.0010) %/Silver (<0.0010) %/Tantalum (<0.0010) %/Arsenic (<0.0050) %/Calcium (<0.0050) %/Cobalt (<0.0050) %/Hafnium (<0.0050) %/Manganese (<0.0050) %/Tungsten (<0.0050) %/Yttrium (<0.0050) %/Boron (<0.0100) %/Lead (<0.0100) %/Magnesium (<0.0100) %/Niobium (<0.0100) %/Zinc (<0.0100) %/Vanadium (0.0017) %/Chromium (0.002) %/Copper (0.003) %/Phosphorus (0.003) %/Oxygen (0.121) %/Titanium (85.5) % |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction