Learn More
7-Hydroxy Memantine Hydrochloride, TRC
Product Code. 30133577
Change view
Click to view available options
Quantity:
5 mg
25 mg
50 mg
Unit Size:
25mg
50mg
5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30133577 50 mg 50mg
30136714 25 mg 25mg
17971695 5 mg 5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 30133577 Supplier Toronto Research Chemicals Supplier No. TRCH94470050MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 356572-08-2
Molecular Formula C12 H21 N O . Cl H
Molecular Weight (g/mol) 231.76
Synonym 3-Amino-5,7-dimethyltricyclo[3.3.1.13,7]decan-1-ol Hydrochloride (1:1), Tricyclo[3.3.1.13,7]decan-1-ol, 3-amino-5,7-dimethyl-, hydrochloride (9CI), 1-Amino-3,5-dimethyl-7-hydroxyadamantane hydrochloride, 7-Hydroxy Memantine hydrochloride, 7-Hydroxymemantine hydrochloride, 3-Amino-5,7-dimethyltricyclo[3.3.1.13,7]decan-1-ol Hydrochloride (1:1), 1-Amino-3,5-dimethyl-7-hydroxyadamantane hydrochloride
IUPAC Name 3-amino-5,7-dimethyladamantan-1-ol;hydrochloride
SMILES Cl.CC12CC3(C)CC(N)(C1)CC(O)(C3)C2
Specifications
Quantity 50 mg
InChI Formula InChI=1S/C12H21NO.ClH/c1-9-3-10(2)5-11(13,4-9)8-12(14,6-9)7-10;/h14H,3-8,13H2,1-2H3;1H
Formula Weight 231.139
Chemical Name or Material 7-Hydroxy Memantine Hydrochloride