missing translation for 'onlineSavingsMsg'
Learn More

7-Methoxy-4-(trifluoromethyl)coumarin, TRC

Product Code. 30479673
Change view
Click to view available options
Quantity:
100 mg
500 mg
1 g
Unit Size:
100mg
1g
500mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30479673 500 mg 500mg
30469482 1 g 1g
30482084 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30479673 Supplier Toronto Research Chemicals Supplier No. TRCM271990500MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 575-04-2
Molecular Formula C8 H8 Br N3
Molecular Weight (g/mol) 226.073
Synonym 7-Methoxy-4-(trifluoromethyl)-2H-1-benzopyran-2-one, 4-(Trifluoromethyl)-7-methoxycoumarin, 7-Methoxy-4-trifluoromethylcoumarin
IUPAC Name 5-bromo-1-methylindazol-3-amine
SMILES Cn1nc(N)c2cc(Br)ccc12

Specifications

Quantity 500 mg
InChI Formula InChI=1S/C8H8BrN3/c1-12-7-3-2-5(9)4-6(7)8(10)11-12/h2-4H,1H3,(H2,10,11)
Formula Weight 224.99
Chemical Name or Material 7-Methoxy-4-(trifluoromethyl)coumarin
Título del producto
Seleccione un problema

Al hacer clic en Enviar, acepta que Fisher Scientific se ponga en contacto con usted en relación con los comentarios que ha proporcionado en este formulario. No compartiremos su información para ningún otro fin. Toda la información de contacto proporcionada se mantendrá de acuerdo con nuestra Política de Privacidad. Política de privacidad.