missing translation for 'onlineSavingsMsg'
Learn More
Learn More
AARSD1 Antibody (Tsp232A), Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-12887-0.025mg
This item is not returnable.
View return policy
Description
AARSD1 Monoclonal specifically detects AARSD1 in Human, Mouse samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin, Microarray.
Specifications
| AARSD1 | |
| Monoclonal | |
| 1 mg/mL | |
| Western Blot 1:100-1:2000, Immunohistochemistry 1:10-1:500, Immunohistochemistry-Paraffin 1:10-1:500 | |
| AARSD1 | |
| Recombinant tsp23 | |
| 0.025 mg | |
| Primary | |
| Detects approx 19kDa. Detects Tsp23, no cross-reactivity to p23. | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. | |
| IgG1 κ |
| Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| Tsp232A | |
| Unconjugated | |
| alanyl-tRNA editing protein Aarsd1, alanyl-tRNA synthetase domain containing 1, Alanyl-tRNA synthetase domain-containing protein 1, MGC2744 | |
| Mouse | |
| Protein G purified | |
| RUO | |
| 80755 | |
| Human, Mouse | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction