missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ABCA6 Antibody - Azide and BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-92497-0.1ml
This item is not returnable.
View return policy
Description
ABCA6 Polyclonal antibody specifically detects ABCA6 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunocytochemistry/ Immunofluorescence
Specifications
| ABCA6 | |
| Polyclonal | |
| Western Blot 1:100 - 1:500, Immunocytochemistry/ Immunofluorescence 1:50 - 1:200 | |
| ABC transporter ABCA6, ATP-binding cassette A6, ATP-binding cassette sub-family A member 6, ATP-binding cassette, sub-family A (ABC1), member 6, EC 2.5.1.19, EC 3.6.3, EC 3.6.3.25, EST155051, FLJ43498 | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 690-828 of human ABCA6 (NP_525023.2). RLKCAGSSMFLKRRWGLGYHLSLHRNEICNPEQITSFITHHIPDAKLKTENKEKLVYTLPLERTNTFPDLFSDLDKCSDQGVTGYDISMSTLNEVFMKLEGQSTIEQDFEQVEMIRDSESLNEMELAHSSFSEMQTAVS | |
| 0.1 mL | |
| Cardiovascular Biology, Endocrinology, Signal Transduction | |
| 23460 | |
| Store at -20°C. Avoid freeze-thaw cycles. | |
| IgG |
| Western Blot, Immunofluorescence | |
| Unconjugated | |
| PBS with 50% glycerol, pH7.3. | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse, Rat | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction