missing translation for 'onlineSavingsMsg'
Learn More

ABCA6 Antibody - Azide and BSA Free, Novus Biologicals™

Rabbit Polyclonal Antibody

€ 190.00 - € 463.00

Specifications

Antigen ABCA6
Dilution Western Blot 1:100 - 1:500, Immunocytochemistry/ Immunofluorescence 1:50 - 1:200
Applications Western Blot, Immunofluorescence
Classification Polyclonal
Conjugate Unconjugated
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
18696962
View Documents
Novus Biologicals
NBP2-92497-0.02ml
0.02 mL
€ 190.00
0.02mL
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
18682282
View Documents
Novus Biologicals
NBP2-92497-0.1ml
0.1 mL
€ 463.00
0.10mL
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

ABCA6 Polyclonal antibody specifically detects ABCA6 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunocytochemistry/ Immunofluorescence
Specifications

Specifications

ABCA6
Western Blot, Immunofluorescence
Unconjugated
Rabbit
Cardiovascular Biology, Endocrinology, Signal Transduction
PBS with 50% glycerol, pH7.3.
23460
IgG
Affinity purified
Western Blot 1:100 - 1:500, Immunocytochemistry/ Immunofluorescence 1:50 - 1:200
Polyclonal
Purified
RUO
Human, Mouse, Rat
ABC transporter ABCA6, ATP-binding cassette A6, ATP-binding cassette sub-family A member 6, ATP-binding cassette, sub-family A (ABC1), member 6, EC 2.5.1.19, EC 3.6.3, EC 3.6.3.25, EST155051, FLJ43498
Recombinant fusion protein containing a sequence corresponding to amino acids 690-828 of human ABCA6 (NP_525023.2). RLKCAGSSMFLKRRWGLGYHLSLHRNEICNPEQITSFITHHIPDAKLKTENKEKLVYTLPLERTNTFPDLFSDLDKCSDQGVTGYDISMSTLNEVFMKLEGQSTIEQDFEQVEMIRDSESLNEMELAHSSFSEMQTAVS
Primary
Store at -20°C. Avoid freeze-thaw cycles.
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.