Learn More
Acebutolol hydrochloride
Product Code. 15465959
Change view
Click to view available options
Quantity:
1 g
5 g
10 g
Unit Size:
10g
1g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15465959 10 g 10g
15445959 1 g 1g
15455959 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 15465959 Supplier Thermo Scientific Alfa Aesar Supplier No. J63790.09
 Restricted Item
Estimated Shipment: 30-12-2026
Add to basket
missing translation for 'validMainframeMsg'
Cardioselective beta-adrenergic blocker | CAS: 34381-68-5 | C18H29ClN2O4 | 372.89 g/mol
Cardioselective beta-adrenergic blocker
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 34381-68-5
Molecular Formula C18H29ClN2O4
Molecular Weight (g/mol) 372.89
MDL Number MFCD00078860
InChI Key KTUFKADDDORSSI-UHFFFAOYNA-N
Synonym acebutolol hydrochloride, acebutolol hcl, n-3-acetyl-4-2-hydroxy-3-isopropylamino propoxy phenyl butyramide hydrochloride, diasectral, wesfalin, 3'-acetyl-4'-2-hydroxy-3-isopropylamino propoxy butyranilide hydrochloride, dl-1-2-acetyl-4-butyramidophenoxy-2-hydroxy-3-isopropylaminopropane hydrochloride, ccris 1102, sectral tn
PubChem CID 441307
ChEBI CHEBI:2380
IUPAC Name hydrogen N-(3-acetyl-4-{2-hydroxy-3-[(propan-2-yl)amino]propoxy}phenyl)butanamide chloride
SMILES [H+].[Cl-].CCCC(=O)NC1=CC=C(OCC(O)CNC(C)C)C(=C1)C(C)=O
Specifications
Melting Point 141°C to 143°C
Color White
Quantity 10 g
Merck Index 14,19
Formula Weight 372.89
Physical Form Powder
Chemical Name or Material Acebutolol hydrochloride