missing translation for 'onlineSavingsMsg'
Learn More

Aceclofenac Ethyl Ester, TRC

Product Code. 18010080
Change view
Click to view available options
Quantity:
50 mg
100 mg
250 mg
Unit Size:
100mg
250mg
50mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18010080 50 mg 50mg
18081250 250 mg 250mg
18054040 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 18010080 Supplier Toronto Research Chemicals Supplier No. TRCA13041550MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 139272-67-6
Molecular Formula C18 H17 Cl2 N O4
Molecular Weight (g/mol) 382.24
Synonym Ethyl [[[2-[(2,6-Dichlorophenyl)amino]phenyl]acetyl]oxy]acetate, Aceclofenac Imp. E (EP), Ethyl Ester of Aceclofenac
IUPAC Name (2-ethoxy-2-oxoethyl) 2-[2-(2,6-dichloroanilino)phenyl]acetate
SMILES CCOC(=O)COC(=O)Cc1ccccc1Nc2c(Cl)cccc2Cl

Specifications

Quantity 50 mg
InChI Formula InChI=1S/C18H17Cl2NO4/c1-2-24-17(23)11-25-16(22)10-12-6-3-4-9-15(12)21-18-13(19)7-5-8-14(18)20/h3-9,21H,2,10-11H2,1H3
Formula Weight 381.0535
Percent Purity >95%
Chemical Name or Material Aceclofenac Ethyl Ester
Grade HPLC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.