Learn More
VELP Scientifica™ Acid Pump Kit for UDK1X9
Product Code. 17382131 Shop All Velp Products
Change view
Click to view available options
For Use With (Equipment):
UDK1X9 NaOH Tank with Caps
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. For Use With (Equipment) unitSize
17382131 UDK1X9 NaOH Tank with Caps Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 17382131 Supplier VELP Scientifica™ Supplier No. A00000422
Only null left
Add to basket
Add to basket
Acid pump for the determination of alcohol, volatile acids, and phenols.
  • Acid pump for use with VELP™ UDK1X9 NaOH Tank with Caps
Specifications
Voltage 230 V
Type Acid Pump
For Use With (Equipment) UDK1X9 NaOH Tank with Caps