Learn More
ACMSD Rabbit, anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand:  Proteintech 26834-1-AP-150UL
 View more versions of this product
Product Code. 18056195
  • € 410.00 / 150µL
Description
The ACMSD antibody from Proteintech is a rabbit polyclonal antibody generated with recombinant protein of human ACMSD. This antibody recognizes human, mouse antigen. The ACMSD antibody has been validated for the following applications: IHC, ELISA analysis.
Specifications
Specifications
ACMSD
400 μg/mL
Rabbit
Primary
Human, Mouse
150 kDa
IgG
Immunohistochemistry, ELISA
ACMSD, Picolinate carboxylase
150 μL
130013
Store at -20°C. Stable for one year after shipment.