Learn More
ACMSD Rabbit, anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 26834-1-AP-150UL
This item is not returnable.
View return policy
Description
The ACMSD antibody from Proteintech is a rabbit polyclonal antibody generated with recombinant protein of human ACMSD. This antibody recognizes human, mouse antigen. The ACMSD antibody has been validated for the following applications: IHC, ELISA analysis.
Specifications
| ACMSD | |
| 400 μg/mL | |
| Rabbit | |
| Primary | |
| Human, Mouse | |
| 150 kDa | |
| IgG |
| Immunohistochemistry, ELISA | |
| ACMSD, Picolinate carboxylase | |
| 150 μL | |
| 130013 | |
| Store at -20°C. Stable for one year after shipment. | |
| Liquid |