Learn More
ActivArmr™ 78-102 Food-Safe Thermal Insulating Gloves with Micro-Dot Palm ActivArmr™ 78-102 Food-Safe Thermal Insulating Gloves with Micro-Dot Palm
Product Code. p-7215026 Shop All Ansell Products
Click to view available options
Size:
7
9
Unit Size:
12 pairs
Product Code. 16825250
Brand: ActivArmr™ 78-102 Food-Safe Thermal Insulating Gloves with Micro-Dot Palm 78202070
Only null left
Add to basket
Add to basket
Seamless knit gloves with a thermal acrylic liner to provide comfort and warmth in cold or refrigerated work environments. Micro-dot coated palm increases abrasion resistance and enhances grip capabilities.
  • Ideal for keeping hands warm in food processing and service applications
  • Seamless knit, thermal acrylic liner provides comfort and warmth in cold or refrigerated work environments
  • Uncoated liner can be used alone, or as an underglove
  • Micro-dot coating on the palm enhances grip capabilities
TRUSTED_SUSTAINABILITY
Specifications
Certifications/Compliance (Europe) EN 420:2003 + A1:2009, EN 511:2006, REACH Compliant
Coating Material PVC
Color Blue
Exterior Finish No Texture
Hand Specific
Material Acrylic
Product Type Thermal Mechanic's Gloves
Size 7
EN388 Standard 312XB
For Use With (Application) Food Processing
Liner Material Acrylic
Series 78-202
Yarn Gauge 10