missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ansell ActivArmr™ 97-125 Cut and Impact Protection Gloves
Shop All Ansell Products
Click to view available options
Size:
8
9
10
11
12
Unit Size:
12 pairs
Description
- HIVIZ™ design increases awareness of users for safety
- Double-dip nitrile coating provides grip and control when working with oils and other liquids
- HPPE seamless knit glove keeps hands cool and comfortable
- Double-dip nitrile coating defends against oil exposure and abrasion injuries
- Designed for dynamic environments which have impact and cut hazards present
Specifications
Specifications
| ANSI Abrasion Rating | ANSI 5 |
| Certifications/Compliance (Europe) | CE, EN 420:2003 + A1:2009; Category II |
| Construction | Knit |
| Coating Material | Nitrile |
| Color | Gray, Hi-vis Orange |
| Cut Resistance Rating | ANSI A4 |
| Product Type | Cut-resistant Gloves |
| Size | 10 |
| EN388 Standard | EN 388:4X43CP |
| For Use With (Application) | Engagement and release of drill string slips, drilling assembly and completion, handling jagged metal, production Services, drilling/extraction, heavy equipment servicing, transport and logistics |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction