missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ADAM8 Antibody (MM0079-6J12) - Azide and BSA Free, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-12020-0.025MG
This item is not returnable.
View return policy
Description
ADAM8 Monoclonal specifically detects ADAM8 in Human samples. It is validated for Western Blot.
Specifications
| ADAM8 | |
| Monoclonal | |
| LYOPH | |
| Western Blot 1:100-1:2000 | |
| P78325 | |
| ADAM8 | |
| Human ADAM8 | |
| 0.025 mg | |
| Cellular Markers | |
| 101 | |
| Human | |
| Purified |
| Western Blot | |
| MM0079-6J12 | |
| Unconjugated | |
| A 0.2 um filtered solution in PBS with No Preservative | |
| a disintegrin and metalloproteinase domain 8, ADAM 8, ADAM metallopeptidase domain 8, CD156a antigen, CD156human leukocyte differentiation antigen, Cell surface antigen MS2, EC 3.4.24, EC 3.4.24.-, MGC134985, MS2disintegrin and metalloproteinase domain-containing protein 8 | |
| Mouse | |
| Protein G purified | |
| RUO | |
| Primary | |
| Reconstitute with 0.2 ml sterilized PBS to a final concentration of 0.5 mg/ml. | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction