missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ADAM8 Antibody (MM0079-6J12) - Azide and BSA Free, Novus Biologicals™
Mouse Monoclonal Antibody
€ 195.00 - € 500.00
Specifications
| Antigen | ADAM8 |
|---|---|
| Clone | MM0079-6J12 |
| Concentration | LYOPH |
| Applications | Western Blot |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18057576
|
Novus Biologicals
NBP2-12020-0.025MG |
0.025 mg |
€ 195.00
25µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18032277
|
Novus Biologicals
NBP2-12020 |
0.1 mg |
€ 500.00
0.10mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
ADAM8 Monoclonal specifically detects ADAM8 in Human samples. It is validated for Western Blot.Specifications
| ADAM8 | |
| LYOPH | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human | |
| a disintegrin and metalloproteinase domain 8, ADAM 8, ADAM metallopeptidase domain 8, CD156a antigen, CD156human leukocyte differentiation antigen, Cell surface antigen MS2, EC 3.4.24, EC 3.4.24.-, MGC134985, MS2disintegrin and metalloproteinase domain-containing protein 8 | |
| ADAM8 | |
| Protein G purified |
| MM0079-6J12 | |
| Western Blot | |
| Unconjugated | |
| Mouse | |
| Cellular Markers | |
| P78325 | |
| 101 | |
| Primary | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title