missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ADAM8 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody has been used in 1 publication
Brand: Novus Biologicals NB600-1393-0.025mg
This item is not returnable.
View return policy
Description
ADAM8 Polyclonal specifically detects ADAM8 in Human, Rat, Porcine samples. It is validated for Western Blot.
Specifications
| ADAM8 | |
| Polyclonal | |
| Unconjugated | |
| PBS and 50% Glycerol with 0.05% Sodium Azide | |
| ADAM8 | |
| Synthetic peptide (Human) corresponding to the amino terminal end of the catalytic domain. | |
| 0.025 mg | |
| Cellular Markers | |
| 101 | |
| Human | |
| IgG |
| Western Blot | |
| 1 mg/mL | |
| Western Blot 1:1000 | |
| a disintegrin and metalloproteinase domain 8, ADAM 8, ADAM metallopeptidase domain 8, CD156a antigen, CD156human leukocyte differentiation antigen, Cell surface antigen MS2, EC 3.4.24, EC 3.4.24.-, MGC134985, MS2disintegrin and metalloproteinase domain-containing protein 8 | |
| Rabbit | |
| Affinity Purified | |
| RUO | |
| Primary | |
| Does not react with other ADAMs. | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze/thaw cycles. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction