missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ADAM8 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody has been used in 1 publication
€ 258.00
Specifications
| Antigen | ADAM8 |
|---|---|
| Concentration | 1 mg/mL |
| Dilution | Western Blot 1:1000 |
| Applications | Western Blot |
| Classification | Polyclonal |
Description
ADAM8 Polyclonal specifically detects ADAM8 in Human, Rat, Porcine samples. It is validated for Western Blot.Specifications
| ADAM8 | |
| Western Blot 1:1000 | |
| Polyclonal | |
| Rabbit | |
| Cellular Markers | |
| PBS and 50% Glycerol with 0.05% Sodium Azide | |
| 101 | |
| Synthetic peptide (Human) corresponding to the amino terminal end of the catalytic domain. | |
| Primary | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze/thaw cycles. |
| 1 mg/mL | |
| Western Blot | |
| Unconjugated | |
| RUO | |
| Human | |
| a disintegrin and metalloproteinase domain 8, ADAM 8, ADAM metallopeptidase domain 8, CD156a antigen, CD156human leukocyte differentiation antigen, Cell surface antigen MS2, EC 3.4.24, EC 3.4.24.-, MGC134985, MS2disintegrin and metalloproteinase domain-containing protein 8 | |
| ADAM8 | |
| IgG | |
| Affinity Purified | |
| Does not react with other ADAMs. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title