Learn More
ANSELL™ Aggressive Chemicals Medium Duty Environment AlphaTec™ Gloves
Product Code. p-200019969 Shop All Ansell Products
Click to view available options
Size:
8
9
10
Unit Size:
1 pair
Product Code. 17963532
Brand: ANSELL™ 55300100
Only null left
Add to basket
Add to basket
AlphaTec™ Neoprene RABS and isolator gloves deliver exceptional chemical resistance against most acids, alcohols, oils, lubricants and hydrocarbons and offer strong physical protection against abrasions and good protection against cuts and punctures.
  • Effective protection for medium duty chemical environments, delivering exceptional chemical resistance against most acids, alcohols, oils, lubricants and hydrocarbons
  • The medium thickness (20 mil or 0.51 mm) increases dexterity, which improves the ability to manipulate objects
  • Unlined for effective cleaning
  • Available port sizes fit most isolator glove boxes
TRUSTED_SUSTAINABILITY
Specifications
Certifications/Compliance (Europe) CE 0493|EN 388:2016 +A1:2018|EN ISO 21420:2020|EN ISO 374-1:2016|EN ISO 374-5:2016|EN 388:2016|EN 420:2003 + A1:2009|Category III|UKCA 0321
Chlorinated No
Color Black
Finger Thickness (English) 20 mil
Finger Thickness (Metric) 0.51 mm
Hand Specific
Latex Free Yes
Material Neoprene Polymer
Palm Thickness (English) 20 mil
Palm Thickness (Metric) 0.51 mm
Product Type Isolator Gloves
Size 10
Thickness (English) 20 mil
Thickness (Metric) 0.51 mm
For Use With (Application) RABS/Isolator Gloves
For Use With (Equipment) RABS/Isolator Gloves
Grip Design Smooth
Port Size (English) 6 in.
Port Size (Metric) 152 mm
Length (English) 28 in.
Length (Metric) 711 mm