missing translation for 'onlineSavingsMsg'
Learn More
Learn More
AHNAK2 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP1-57661
This item is not returnable.
View return policy
Description
AHNAK2 Polyclonal specifically detects AHNAK2 in Human samples. It is validated for Western Blot.
Specifications
| AHNAK2 | |
| Polyclonal | |
| Unconjugated | |
| PBS, 2% Sucrose with 0.09% Sodium Azide | |
| AHNAK2 | |
| The immunogen is a synthetic peptide directed towards the middle region of human AHNAK2. Peptide Sequence KEEGELIGPVGTGLDSRVMVTSAARTELILPEQDRKADDESKGSGLGPNEG. The peptide sequence for this immunogen was taken from within the described region. | |
| 100 μL | |
| Primary | |
| Expected identity based on immunogen sequence: Human: 100%. | |
| Human, Yeast | |
| IgG |
| Western Blot | |
| 0.5 mg/ml | |
| Western Blot 1.0 ug/ml | |
| AHNAK nucleoprotein 2, C14orf78, chromosome 14 open reading frame 78, KIAA2019, protein AHNAK2 | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| 113146 | |
| Centrifuge the vial of lyoph antibody at 12,000 x g for 20 seconds. Add 50μL of distilled water. Vortex followed by centrifuge again to pellet the solution.Final concentration is 1mg/mL in PBS buffer. | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction