missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Anti-Rat CD45RA (OX-33), Proteintech
Mouse Monoclonal Antibody
Brand: Proteintech 65234-1-IG-500UG
This item is not returnable.
View return policy
Description
The CD45RA antibody from Proteintech is a mouse monoclonal antibody generated with leukocyte common antigen purified from rat splenocytes of rat CD45RA. This antibody recognizes rat antigen. The CD45RA antibody has been validated for the following applications: FC analysis.Specifications
| CD45RA | |
| Monoclonal | |
| 0.5 mg/mL | |
| Mouse | |
| Affinity purification | |
| RUO | |
| 24699 | |
| Store at 2-8°C. Stable for one year after shipment. | |
| Liquid |
| Flow Cytometry | |
| OX-33 | |
| Unconjugated | |
| Leukocyte common antigen purified from rat splenocytes | |
| 500 μg | |
| Primary | |
| Rat | |
| Flow Antibody | |
| IgG1 κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction