missing translation for 'onlineSavingsMsg'
Learn More
Learn More
AHSA1 Antibody (4H9-D7), Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-12891-0.025mg
This item is not returnable.
View return policy
Description
AHSA1 Monoclonal specifically detects AHSA1 in Human, Mouse samples. It is validated for Western Blot, ELISA, Immunocytochemistry/Immunofluorescence.
Specifications
| AHSA1 | |
| Monoclonal | |
| 1 mg/mL | |
| Western Blot 1:100-1:2000, ELISA 1:100-1:2000, Immunocytochemistry/Immunofluorescence | |
| activator of 90 kDa heat shock protein ATPase homolog 1, AHA1, activator of heat shock 90kDa protein ATPase homolog 1 (yeast), C14orf3, chromosome 14 open reading frame 3, p38AHA1 | |
| Mouse | |
| Protein G purified | |
| RUO | |
| 10598 | |
| Human, Mouse | |
| Purified |
| Western Blot, ELISA, Immunocytochemistry, Immunofluorescence | |
| 4H9-D7 | |
| Unconjugated | |
| NP_036243.1 | |
| AHSA1 | |
| Human Aha1, his tagged | |
| 0.025 mg | |
| Primary | |
| Detects approx 38kDa, can run up to 45kDa on SDS Page. It sees human Aha1, Mouse Aha1 and Mouse Aha2. | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. | |
| IgG1 κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction