missing translation for 'onlineSavingsMsg'
Learn More
Learn More
AHSA1 Antibody (4H9-D7), Novus Biologicals™
Mouse Monoclonal Antibody
€ 186.00
Specifications
| Antigen | AHSA1 |
|---|---|
| Clone | 4H9-D7 |
| Concentration | 1 mg/mL |
| Dilution | Western Blot 1:100-1:2000, ELISA 1:100-1:2000, Immunocytochemistry/Immunofluorescence |
| Applications | Western Blot, ELISA, Immunocytochemistry, Immunofluorescence |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18742994
|
Novus Biologicals
NBP2-12891-0.025mg |
0.025 mg | |||||||
Description
AHSA1 Monoclonal specifically detects AHSA1 in Human, Mouse samples. It is validated for Western Blot, ELISA, Immunocytochemistry/Immunofluorescence.Specifications
| AHSA1 | |
| 1 mg/mL | |
| Western Blot, ELISA, Immunocytochemistry, Immunofluorescence | |
| Unconjugated | |
| Mouse | |
| Human, Mouse | |
| activator of 90 kDa heat shock protein ATPase homolog 1, AHA1, activator of heat shock 90kDa protein ATPase homolog 1 (yeast), C14orf3, chromosome 14 open reading frame 3, p38AHA1 | |
| AHSA1 | |
| IgG1 κ | |
| Protein G purified | |
| Detects approx 38kDa, can run up to 45kDa on SDS Page. It sees human Aha1, Mouse Aha1 and Mouse Aha2. |
| 4H9-D7 | |
| Western Blot 1:100-1:2000, ELISA 1:100-1:2000, Immunocytochemistry/Immunofluorescence | |
| Monoclonal | |
| Purified | |
| RUO | |
| NP_036243.1 | |
| 10598 | |
| Human Aha1, his tagged | |
| Primary | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title