missing translation for 'onlineSavingsMsg'
Learn More
Learn More
AHSA1 Mouse anti-Human, Clone: 3.00E+09, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-59405-50UL
This item is not returnable.
View return policy
Description
AHSA1 Monoclonal specifically detects AHSA1 in Human samples. It is validated for Western Blot, Flow Cytometry, ELISA, Immunocytochemistry/Immunofluorescence.
Specifications
| AHSA1 | |
| Monoclonal | |
| 1 mg/mL | |
| Western Blot 1:1000, Flow Cytometry 1:10-1:1016, ELISA, Immunocytochemistry/Immunofluorescence 1:100 | |
| activator of 90 kDa heat shock protein ATPase homolog 1, AHA1, activator of heat shock 90kDa protein ATPase homolog 1 (yeast), C14orf3, chromosome 14 open reading frame 3, p38AHA1 | |
| Mouse | |
| Protein A purified | |
| RUO | |
| 10598 | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. | |
| IgG2b κ |
| Western Blot, Flow Cytometry, ELISA, Immunocytochemistry, Immunofluorescence | |
| 3000000000 | |
| Unconjugated | |
| Liquid. In PBS (pH 7.4), 10% Glycerol with 0.02% Sodium Azide | |
| AHSA1 | |
| Recombinant human AHA1(19-337aa) purified from E. coli | |
| 50 μL | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction