missing translation for 'onlineSavingsMsg'
Learn More
Learn More
AHSA1 Mouse anti-Human, Clone: 3.00E+09, Novus Biologicals™
Mouse Monoclonal Antibody
€ 315.00 - € 509.00
Specifications
| Antigen | AHSA1 |
|---|---|
| Clone | 3000000000 |
| Concentration | 1 mg/mL |
| Dilution | Western Blot 1:1000, Flow Cytometry 1:10-1:1016, ELISA, Immunocytochemistry/Immunofluorescence 1:100 |
| Applications | Western Blot, Flow Cytometry, ELISA, Immunocytochemistry, Immunofluorescence |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18215843
|
Novus Biologicals
NBP2-59405-50UL |
50 μL |
€ 315.00
50µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18236254
|
Novus Biologicals
NBP2-59405-100UL |
100 μL |
€ 509.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
AHSA1 Monoclonal specifically detects AHSA1 in Human samples. It is validated for Western Blot, Flow Cytometry, ELISA, Immunocytochemistry/Immunofluorescence.Specifications
| AHSA1 | |
| 1 mg/mL | |
| Western Blot, Flow Cytometry, ELISA, Immunocytochemistry, Immunofluorescence | |
| Unconjugated | |
| Mouse | |
| Human | |
| activator of 90 kDa heat shock protein ATPase homolog 1, AHA1, activator of heat shock 90kDa protein ATPase homolog 1 (yeast), C14orf3, chromosome 14 open reading frame 3, p38AHA1 | |
| AHSA1 | |
| IgG2b κ | |
| Protein A purified |
| 3000000000 | |
| Western Blot 1:1000, Flow Cytometry 1:10-1:1016, ELISA, Immunocytochemistry/Immunofluorescence 1:100 | |
| Monoclonal | |
| Purified | |
| RUO | |
| Liquid. In PBS (pH 7.4), 10% Glycerol with 0.02% Sodium Azide | |
| 10598 | |
| Recombinant human AHA1(19-337aa) purified from E. coli | |
| Primary | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title