missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ AK1BA Polyclonal Antibody

Rabbit Polyclonal Antibody

Brand:  Invitrogen™ PA5122255

 View more versions of this product

Product Code. 17733654

  • € 208.00 / 20µL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

AK1BA Polyclonal Antibody for Western Blot, ICC/IF, IHC

Catalyzes the NADPH-dependent reduction of a wide variety of carbonyl-containing compounds to their corresponding alcohols. Displays strong enzymatic activity toward all-trans-retinal, 9-cis-retinal, and 13-cis-retinal. Plays a critical role in detoxifying dietary and lipid-derived unsaturated carbonyls, such as crotonaldehyde, 4-hydroxynonenal, trans-2-hexenal, trans-2,4-hexadienal and their glutathione-conjugates carbonyls (GS-carbonyls). Displays no reductase activity towards glucose.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

AK1BA
Polyclonal
Unconjugated
AKR1B10
AKR1B10; AKR1B11; Aldo-keto reductase family 1 member B10
Rabbit
Antigen affinity chromatography, Protein A
RUO
296972, 57016
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Liquid
Immunohistochemistry, Western Blot, Immunocytochemistry
0.5 mg/mL
PBS with 50% glycerol and 0.05% ProClin 300; pH 7.4
O60218
AKR1B10
Recombinant protein Aldose Reductase Like Protein 1. The antigen corresponds to amino acid range 1-316 of the target protein.
20 μL
Primary
Human, Rat
Antibody
IgG
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.