missing translation for 'onlineSavingsMsg'
Learn More
Learn More
AKT1, p Ser129 Antibody (SR2047), Novus Biologicals™
Rabbit Monoclonal Antibody
€ 338.00 - € 449.00
Specifications
| Antigen | AKT1 |
|---|---|
| Clone | SR2047 |
| Dilution | Western Blot 1:1000-1:2000 |
| Applications | Western Blot |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18684245
|
Novus Biologicals
NBP3-21567-50ul |
50 μg |
€ 338.00
50µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18632844
|
Novus Biologicals
NBP3-21567-100ul |
100 μg |
€ 449.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
AKT1 Monoclonal antibody specifically detects AKT1 in Human, Mouse, Rat samples. It is validated for Western BlotSpecifications
| AKT1 | |
| Western Blot 1:1000-1:2000 | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human, Mouse, Rat | |
| AKT, AKT serine/threonine kinase 1, AKT1m, CWS6, EC 2.7.11, EC 2.7.11.1, PKB, PKB alpha, PKBMGC99656, PRKBA, Protein kinase B, Protein Kinase B Alpha, Proto-oncogene c-Akt, RAC, rac protein kinase alpha, RAC-ALPHA, RAC-alpha serine/threonine-protein kinase, RAC-PK-alpha, RACPKB-ALPHA, Serine-Threonine Protein Kinase, v-akt murine thymoma viral oncogene homolog 1, V-Akt Murine Thymoma Viral Oncogene-Like Protein 1 | |
| A synthesized peptide derived from human phospho-AKT1 [Ser129] (Uniprot #: P31749) | |
| Primary | |
| Store at -20 degrees C. Avoid freeze/thaw cycles. |
| SR2047 | |
| Western Blot | |
| Unconjugated | |
| Rabbit | |
| Angiogenesis, Apoptosis, Autophagy, Cancer, Cellular Markers, Growth and Development, Hypoxia, mTOR Pathway, Neuronal Cell Markers, Phospho Specific, Protein Kinase, Serine Kinases, Signal Transduction, Tyrosine Kinases | |
| PBS, pH 7.4, 150mM NaCl, 50% glycerol. | |
| 207 | |
| IgG | |
| Affinity purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title