Learn More
Al-23 Multibore Round Tube, O.D.: 5mm, Four 1mm Bores, Thermo Scientific™
Product Code. 11352948
Click to view available options
Quantity:
100 mm
200 mm
250 mm
300 mm
600 mm
800 mm
1000 mm
Unit Size:
1000mm
100mm
200mm
250mm
300mm
600mm
800mm
Product Code. 11352948
Brand: Thermo Scientific Alfa Aesar 032710.EH
Request Bulk or Custom Format
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
TRUSTED_SUSTAINABILITY
Specifications
Chemical Name or Material Al-23 Multibore Round Tube
Name Note Four bores
Quantity 600 mm