Learn More
Al-23 Tube (one end closed);OD (mm), 30;ID (mm), 25
Product Code. 11352808
missing translation for 'changeView'
missing translation for 'orderingAttributeHoverText'
Quantity:
100 mm
200 mm
300 mm
600 mm
800 mm
missing translation for 'unitSize'
100mm
200mm
300mm
600mm
800mm
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11352808 100 mm 100mm
11322808 200 mm 200mm
11342808 300 mm 300mm
11332808 600 mm 600mm
11302808 800 mm 800mm
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 missing translation for 'options'
Product Code. 11352808 missing translation for 'mfr' Thermo Scientific Alfa Aesar missing translation for 'supplierNo' 030080.DM
Request Bulk or Custom Format
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
TRUSTED_SUSTAINABILITY
Specifications
Chemical Name or Material Al-23 Tube
Name Note One end closed
Quantity 100 mm
What is the maximum working temperature of Al-23 Tubes?
The maximum working temperature for the Al-23 Tube is 1950 degrees C.