missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Fibrinogen gamma chain Antibody (5A6), Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-37560-0.025ml
This item is not returnable.
View return policy
Description
Fibrinogen gamma chain Monoclonal specifically detects Fibrinogen gamma chain in Human samples. It is validated for Western Blot, ELISA, Immunocytochemistry/Immunofluorescence.
Specifications
| Fibrinogen gamma chain | |
| Monoclonal | |
| Unconjugated | |
| Ascites with 0.03% Sodium Azide | |
| FGG | |
| Purified recombinant fragment of human FGG expressed in E. Coli. | |
| Unpurified | |
| RUO | |
| Primary | |
| Human | |
| Ascites |
| Western Blot, ELISA, Immunocytochemistry, Immunofluorescence | |
| 5A6 | |
| Western Blot 1:500 - 1:2000, ELISA 1:10000, Immunocytochemistry/Immunofluorescence 1:200 - 1:1000 | |
| fibrinogen gamma chain, fibrinogen, gamma polypeptide | |
| Mouse | |
| 52 kDa | |
| 0.025 mL | |
| Angiogenesis, Cancer | |
| 2266 | |
| Store at 4°C short term. Aliquot and store at-20°C long term. Avoid freeze/thaw cycles. | |
| IgG2b |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction