missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Alginate Antibody (3G4-1F5), Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-59349-25ug
This item is not returnable.
View return policy
Description
Alginate Monoclonal specifically detects Alginate in Non-species specific samples. It is validated for Western Blot, Immunohistochemistry.
Specifications
| Alginate | |
| Monoclonal | |
| Unconjugated | |
| PBS (pH 7.4), 50% glycerol with 0.09% Sodium Azide | |
| Mouse | |
| Protein G purified | |
| RUO | |
| Binds selectively to a BSA-conjugated alginate, but not to unconjugated BSA. | |
| Purified |
| Western Blot, Immunohistochemistry | |
| 3G4-1F5 | |
| Western Blot 1:100 - 1:2000, Immunohistochemistry 1:10 - 1:500 | |
| Gum levan, Kelacid, Levan gum, Sazio | |
| Sodium Alginate conjugated to KLH | |
| 25 μg | |
| Primary | |
| Store at -20°C. | |
| IgG1 κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction