missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Alginate Antibody (3G4-1F5), Novus Biologicals™
Mouse Monoclonal Antibody
€ 180.00 - € 559.00
Specifications
| Antigen | Alginate |
|---|---|
| Clone | 3G4-1F5 |
| Dilution | Western Blot 1:100 - 1:2000, Immunohistochemistry 1:10 - 1:500 |
| Applications | Western Blot, Immunohistochemistry |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18074096
|
Novus Biologicals
NBP2-59349 |
100 μg |
€ 559.00
100µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18666647
|
Novus Biologicals
NBP2-59349-25ug |
25 μg |
€ 180.00
25µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Alginate Monoclonal specifically detects Alginate in Non-species specific samples. It is validated for Western Blot, Immunohistochemistry.Specifications
| Alginate | |
| Western Blot 1:100 - 1:2000, Immunohistochemistry 1:10 - 1:500 | |
| Monoclonal | |
| Purified | |
| RUO | |
| Sodium Alginate conjugated to KLH | |
| Primary | |
| Binds selectively to a BSA-conjugated alginate, but not to unconjugated BSA. |
| 3G4-1F5 | |
| Western Blot, Immunohistochemistry | |
| Unconjugated | |
| Mouse | |
| Gum levan, Kelacid, Levan gum, Sazio | |
| IgG1 κ | |
| Protein G purified |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title