missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Alloy Steel, 50g, ARMI∣MBH
Click to view available options
Quantity:
50 g
Unit Size:
50g
Description
Alloy Steel, 50g
Specifications
Specifications
| Chemical Name or Material | Magnesium/Antimony/Lead/Zirconium/Niobium/Tungsten/Zinc/Arsenic/Tin/Sulfur/Phosphorus/Titanium/Copper/Cobalt/Aluminium/Molybdenum/Chromium/Nickel/Silicon/Vanadium/Carbon/Manganese/Boron/Nitrogen/Iron |
| CAS | 7439-89-6 |
| Quantity | 50 g |
| Form | Pieces |
| Concentration or Composition (by Analyte or Components) | Magnesium -100 ppm/Antimony -10 ppm/Lead -10 ppm/Zirconium 0.0013 %/Niobium 0.0039 %/Tungsten 0.004 %/Zinc 0.005 %/Arsenic 0.0101 %/Tin 0.0142 %/Sulfur 0.016 %/Phosphorus 0.04 %/Titanium 0.08 %/Copper 0.107 %/Cobalt 0.151 %/Aluminium 0.177 %/Molybdenum 0.206 %/Chromium 0.291 %/Nickel 0.409 %/Silicon 0.53 %/Vanadium 0.603 %/Carbon 0.681 %/Manganese 0.855 %/Boron (<10) ppm/Nitrogen (<100) ppm/Iron (<95.6) % |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction