Learn More
Alloy Steel, Cr/Mo, AISI 4130, G41300, 50g, ARMI∣MBH
Click to view available options
Quantity:
50 g
Unit Size:
50g
Description
Alloy Steel, Cr/Mo, AISI 4130, G41300, 50g
Specifications
Specifications
| Chemical Name or Material | Aluminium, Arsenic, Carbon, Chromium, Copper, Manganese, Molybdenum, Nickel, Nitrogen, Phosphorus, Silicon, Tin, Zinc |
| Quantity | 50 g |
| Weight | 50 g |
| Form | Pieces |
| Concentration or Composition (by Analyte or Components) | Zinc: 0.0012%, Arsenic: 0.0043%, Tin: 0.006%, Phosphorus: 0.0082%, Nitrogen: 0.0095%, Aluminium: 0.027%, Nickel: 0.084%, Copper: 0.131%, Silicon: 0.183%, Molybdenum: 0.217%, Carbon: 0.319%, Manganese: 0.551%, Chromium: 0.996% |