missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Alloy Steel, Ni/Mo, AISI 4620, G46200, 100g, ARMI∣MBH
Click to view available options
Quantity:
100 g
Unit Size:
100g
Description
Alloy Steel, Ni/Mo, AISI 4620, G46200, 100g
Specifications
Specifications
| Chemical Name or Material | Aluminium/Arsenic/Boron/Calcium/Carbon/Chromium/Cobalt/Copper/Lead/Manganese/Molybdenum/Nickel/Niobium/Nitrogen/Oxygen/Phosphorus/Silicon/Tin/Titanium/Tungsten/Vanadium |
| CAS | 7439-89-6 |
| Quantity | 100 g |
| Form | Pieces |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction