missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Alloy Steel, Reinforcing Steel, A706 Grade 80, ARMI∣MBH
Click to view available options
Unit Size:
Each
Description
Alloy Steel, Reinforcing Steel, A706 Grade 80, Ø32x19mm
Specifications
Specifications
| Chemical Name or Material | Niobium/Titanium/Aluminium/Tungsten/Arsenic/Cobalt/Phosphorus/Tin/Nitrogen/Vanadium/Molybdenum/Chromium/Nickel/Silicon/Carbon/Copper/Manganese/Bismuth/Cadmium/Magnesium/Selenium/Zirconium/Zinc/Tantalum/Lead/Calcium/Boron/Antimony/Oxygen |
| CAS | 7439-89-6 |
| Form | Disc |
| Concentration or Composition (by Analyte or Components) | Niobium 0.0018 %/Titanium 0.002 %/Aluminium 0.0023 %/Tungsten 0.003 %/Arsenic 0.0055 %/Cobalt 0.011 %/Phosphorus 0.013 %/Tin 0.0137 %/Nitrogen 0.014 %/Vanadium 0.0299 %/Molybdenum 0.0341 %/Chromium 0.129 %/Nickel 0.185 %/Silicon 0.269 %/Carbon 0.272 %/Copper 0.379 %/Manganese 1.16 %/Bismuth (<0.0010) %/Cadmium (<0.0010) %/Magnesium (<0.0010) %/Selenium (<0.0010) %/Zirconium (<0.0020) %/Zinc (<0.0030) %/Tantalum (<0.0050) %/Lead (<0.0070) %/Calcium (0.0004) %/Boron (0.0007) %/Antimony (0.005) %/Oxygen (0.005) % |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction