missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Alloy Steel, Resulfurized, 50g, ARMI∣MBH
Click to view available options
Quantity:
50 g
Unit Size:
50g
Description
Alloy Steel, Resulfurized, 50g
Specifications
Specifications
| Chemical Name or Material | Vanadium/Nitrogen/Tin/Cobalt/Phosphorus/Copper/Carbon/Nickel/Molybdenum/Silicon/Chromium/Manganese/Aluminium |
| CAS | 7439-89-6 |
| Quantity | 50 g |
| Form | Pieces |
| Concentration or Composition (by Analyte or Components) | Vanadium 0.0154 %/Nitrogen 0.026 %/Tin 0.0289 %/Cobalt 0.0509 %/Phosphorus 0.0689 %/Copper 0.1073 %/Carbon 0.143 %/Nickel 0.147 %/Molybdenum 0.264 %/Silicon 0.404 %/Chromium 0.89 %/Manganese 1.155 %/Aluminium (0.08) % |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction