missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Copper, Brass Majors, ARMI∣MBH
Click to view available options
Quantity:
1 Ea.
Unit Size:
Each
Description
Copper, Brass Majors, Ø38x15
Specifications
Specifications
| Chemical Name or Material | Cobalt/Selenium/Chromium/Tellurium/Silver/Cadmium/Aluminium/Silicon/Phosphorus/Bismuth/Manganese/Arsenic/Nickel/Antimony/Tin/Iron/Lead/Zinc/Copper/Boron/Sulfur |
| CAS | 7440-50-8 |
| Quantity | 1 Ea. |
| Form | Disc |
| Concentration or Composition (by Analyte or Components) | Cobalt 0.0007 %/Selenium 0.001 %/Chromium 0.0012 %/Tellurium 0.0012 %/Silver 0.0041 %/Cadmium 0.011 %/Aluminium 0.015 %/Silicon 0.015 %/Phosphorus 0.016 %/Bismuth 0.019 %/Manganese 0.0194 %/Arsenic 0.0214 %/Nickel 0.0249 %/Antimony 0.036 %/Tin 0.0556 %/Iron 0.357 %/Lead 2.36 %/Zinc 37.3 %/Copper 59.7 %/Boron (<0.0003) %/Sulfur (0.003) % |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction