Learn More
Alodane, TRC
Product Code. 30133451
Change view
Click to view available options
Quantity:
25 mg
50 mg
100 mg
Unit Size:
100mg
25mg
50mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30133451 100 mg 100mg
30138187 50 mg 50mg
30004495 25 mg 25mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 30133451 Supplier Toronto Research Chemicals Supplier No. TRCA614283100MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 2550-75-6
Molecular Formula C9 H6 Cl8
Molecular Weight (g/mol) 397.77
Synonym 2-Norbornene, 1,2,3,4,7,7-hexachloro-5,6-bis(chloromethyl)- (6CI,7CI,8CI), 1,2,3,4,7,7-Hexachloro-5,6-bis(chloromethyl)bicyclo[2.2.1]hept-2-ene, 1,2,3,4,7,7-Hexachloro-5,6-bis(chloromethyl)-2-norbornene, 1,2,3,4,7,7-Hexachloro-5,6-di(chloromethyl)[2.2.1]bicyclohept-2-ene, AC 12402, Alodan, Alodan (pesticide), Chlorbicyclen, Cyclodan, Hercules 12402, Hercules 426, NSC 41891
IUPAC Name (1R,4S)-1,2,3,4,7,7-hexachloro-5,6-bis(chloromethyl)bicyclo[2.2.1]hept-2-ene
SMILES ClCC1C(CCl)[C@]2(Cl)C(=C(Cl)[C@@]1(Cl)C2(Cl)Cl)Cl
Specifications
Analyte or Component Names Chlorbicyclen
Shipping Condition Room Temperature
InChI Formula InChI=1S/C9H6Cl8/c10-1-3-4(2-11)8(15)6(13)5(12)7(3,14)9(8,16)17/h3-4H,1-2H2/t3?,4?,7-,8+
Solution Type Neat
Industry Type Pharmaceutical
Quantity 100 mg
Formula Weight 393.7978
Chemical Name or Material Chlorbicyclen