Learn More
Ansell AlphaTec™ 87-104 Heavy-Duty Latex Chemical Protection Gloves
Product Code. 17248466 Shop All Ansell Products
Click to view available options
Size:
7.5
8.5
9.5
10.5
Unit Size:
12 pairs
Product Code. 17248466
Brand: Ansell 87104105
Only null left
Add to basket
Add to basket
Gloves offer a high resistance to water-based chemicals, along with flexibility and elasticity to prevent tears.
  • Heavy duty chemical protection gloves constructed of natural rubber latex
  • High resistance to water-based chemicals
  • Chlorinated to harden and cleanse the surface of the glove
  • Elastic and flexible to prevent tears
  • Beaded cuff for easy donning/doffing
TRUSTED_SUSTAINABILITY
Specifications
Certifications/Compliance (Europe) EN 420:2003 + A1:2009; Category III
Applications Industrial
AQL 0.65
Color Black
Exterior Finish No Texture
Latex Free No
Material Latex
Palm Thickness (English) 0.0472 in.
Palm Thickness (Metric) 1.2 mm
Product Type Chemical Protection Gloves
Size 10.5
Thickness (English) 47 mil
Thickness (Metric) 1.2 mm
EN388 Standard 3121B
EN ISO 374-1 Type A
EN ISO 374-1 CHEMICAL LETTER CODES AKLMPT
EN ISO 374 - 5 Bacteria, Fungi only
Glove Cuff Style Beaded
Series AlphaTec™ 87-104
Length (English) 17 in.
Length (Metric) 43.2 cm