Learn More
Lovibond™ Aluminum #2 tablets
ECR method for comparators and photometers
Brand: Lovibond™ 515471BT
Additional Details : CAS : 100-97-0 Weight : 0.06300kg Transport : UN number : 1328 Chem class : 4.1 Pack group : III
Description
ECR method for comparators and photometers
Water analysis couldn't be easier. Precise dosing, long shelf life and easy handling make the tablet reagent the most popular indicator system and the cheapest way to have analyse a parameter.
The tablet reagent in the aluminum blister pack withstands almost all climatic conditions and external influences. In addition, there is no safety risk for the user.
Specifications
250 Tablets | |
Aluminum #2 0.01 to 0.30 mg/L | |
Reagent |
Safety and Handling
- Aluminium No. 2
Signal Word
- Warning
Hazard Category
- Flammable solid Category 2
- Skin sensitiser Category 1
Hazard Statement
- H228-Flammable solid.
- H317-May cause an allergic skin reaction.
Precautionary Statement
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P302+P352-IF ON SKIN: Wash with plenty of water/.
- P333+P313-If skin irritation or rash occurs: Get medical advice/attention.
Supplemental information
- MIXTURE LIST-Contains : methenamine