missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Aluminum, Al/Cu, AA 2024, A92024, 100g, ARMI∣MBH
Click to view available options
Quantity:
100 g
Unit Size:
100g
Description
Aluminum, Al/Cu, AA 2024, A92024, 100g
Specifications
Specifications
| Chemical Name or Material | Beryllium/Vanadium/Zirconium/Bismuth/Chromium/Zinc/Tin/Lead/Gallium/Nickel/Titanium/Iron/Silicon/Manganese/Magnesium/Copper/Boron/Aluminium/Phosphorus/Calcium |
| CAS | 7429-90-5 |
| Quantity | 100 g |
| Form | Pieces |
| Concentration or Composition (by Analyte or Components) | Beryllium 0.004 %/Vanadium 0.01 %/Zirconium 0.012 %/Bismuth 0.015 %/Chromium 0.016 %/Zinc 0.017 %/Tin 0.02 %/Lead 0.025 %/Gallium 0.026 %/Nickel 0.03 %/Titanium 0.03 %/Iron 0.25 %/Silicon 0.31 %/Manganese 0.65 %/Magnesium 1.65 %/Copper 4.66 %/Boron (0.002) %/Aluminium (92.27) %/Phosphorus [0.002] %/Calcium [0.007] % |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction