missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Aluminum, Al/Si, ARMI∣MBH
Click to view available options
Quantity:
1 Ea.
Unit Size:
Each
Description
Aluminum, Al/Si, Ø50x17mm
Specifications
Specifications
| CAS | 7429-90-5 |
| Quantity | 1 Ea. |
| Form | Disc |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction