missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Aluminum, Al/Zn/Mg/Cu, AA 7022, A97022, ARMI∣MBH
Click to view available options
Quantity:
1 Ea.
Unit Size:
Each
Description
Aluminum, Al/Zn/Mg/Cu, AA 7022, A97022, Ø50x20mm
Specifications
Specifications
| Chemical Name or Material | Tin/Boron/Zirconium/Lead/Nickel/Titanium/Iron/Silicon/Manganese/Chromium/Copper/Magnesium/Zinc |
| CAS | 7429-90-5 |
| Quantity | 1 Ea. |
| Form | Disc |
| Concentration or Composition (by Analyte or Components) | Tin 0.0006 %/Boron 0.0015 %/Zirconium 0.0016 %/Lead 0.0019 %/Nickel 0.003 %/Titanium 0.016 %/Iron 0.12 %/Silicon 0.126 %/Manganese 0.127 %/Chromium 0.14 %/Copper 0.631 %/Magnesium 2.69 %/Zinc 5.17 % |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction