missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Aluminum, Al/Zn/Mg/Cu, AA 7068, A97068, ARMI∣MBH
Click to view available options
Quantity:
1 Ea.
Unit Size:
Each
Description
Aluminum, Al/Zn/Mg/Cu, AA 7068, A97068, Ø50x20mm
Specifications
Specifications
| Chemical Name or Material | Lead/Nickel/Manganese/Chromium/Titanium/Silicon/Zirconium/Iron/Copper/Magnesium/Zinc/Tin |
| CAS | 7429-90-5 |
| Quantity | 1 Ea. |
| Form | Disc |
| Concentration or Composition (by Analyte or Components) | Lead 0.002 %/Nickel 0.0056 %/Manganese 0.0156 %/Chromium 0.0324 %/Titanium 0.0419 %/Silicon 0.051 %/Zirconium 0.088 %/Iron 0.117 %/Copper 1.676 %/Magnesium 2.7 %/Zinc 7.96 %/Tin (0.0007) % |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction