Learn More
Aluminum oxide, Aerosol Refractory Brushable Paint
Product Code. p-7058722
Click to view available options
Quantity:
250 g
1 kg
Unit Size:
1kg
250g
Product Code. 11364788
Brand: Thermo Scientific Alfa Aesar 043119.30
Request Bulk or Custom Format
Provides protective high temperature Al2O3 coating on ceramic, graphite, or metal | CAS: 1344-28-1 | Al2O3 | 101.96 g/mol
Provides protective high temperature Al{2}O{3} coating on ceramic, graphite, or metalAluminum oxide, Aerosol Refractory Brushable Paint is used as barrier layer between dissimilar materials to prevent reaction and interactions during high temperature. It also acts as a solvent-based paint. It also provides protective high temperature alumina coating on ceramic, graphite and metal.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 1344-28-1
Molecular Formula Al2O3
Molecular Weight (g/mol) 101.96
MDL Number MFCD00003424
InChI Key PNEYBMLMFCGWSK-UHFFFAOYSA-N
Synonym aluminum oxide, alpha-alumina, fasertonerde, abramant, abramax, abrarex, abrasit, aloxite, alundum
PubChem CID 9989226
SMILES [O--].[O--].[O--].[Al+3].[Al+3]
Specifications
Color White
Physical Form Paste
Quantity 250 g
UN Number UN1263
Flash Point −4°C (25°F)
Solubility Information Slightly soluble in water.
IUPAC Name dialuminium(3+) trioxidandiide
Chemical Name or Material Aluminum oxide, Aerosol Refractory Brushable Paint
compliance-icons compliance-icons
Product Identifier
  • Aluminum oxide, Aerosol Refractory Brushable Paint
Signal Word
  • Danger
Hazard Category
  • Flammable liquid Category 2
  • Serious eye damage/eye irritation Category 2
Hazard Statement
  • H225-Highly flammable liquid and vapour.
  • H319-Causes serious eye irritation.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P403+P235-Store in a well-ventilated place. Keep cool.
  • P501-Dispose of contents/container to .
Supplemental information
  • MIXTURE LIST-Contains : Ethyl alcohol, Acetone, Hectorite
For Manufacturing and Laboratory Use. Not for beverage use