missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ammonium Chloride, MP Biomedicals™
Shop All MP Biomedicals Products
Click to view available options
Quantity:
500 g
Unit Size:
500g
Description
Industrial applications include as a flux for coating sheet iron with zinc; tinning; in dry and Leclanch batteries; dyeing, freezing mixtures, electroplating, to clean soldering irons, safety explosives, lustering cotton, tanning; in washing powders; manufacturing of dyes; in cement for iron pipes; for snow treatment (slows melting on ski slopes).
Specifications
Specifications
| CAS | 12125-02-9 |
| Physical Form | Powder |
| Quantity | 500 g |
| Molecular Formula | ClH4N |
| MDL Number | MFCD00011420 |
| Synonym | ammonium chloride, salmiac, sal ammoniac, ammonium muriate, ammoniumchlorid, ammoneric, darammon, amchlor, ammonium chloride nh4 cl, ammoniumklorid |
| Solubility Information | Soluble in water (100mg/mL, clear, colorless solution). The presence of hydrochloric acid or sodium chloride decrease water solubility. Also soluble in methanol and ethanol. |
| InChI Key | NLXLAEXVIDQMFP-UHFFFAOYSA-N |
| SMILES | N.Cl |
| IUPAC Name | amine hydrochloride |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction