missing translation for 'onlineSavingsMsg'
Learn More

Ammonium oxalate monohydrate, ACS, 99.0-101.0%

Product Code. 11383278
Change view
Click to view available options
Quantity:
100 g
500 g
4 x 500 g
Unit Size:
100g
2000g
500g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11383278 500 g 500g
11393278 100 g 100g
11373278 4 x 500 g 2000g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11383278 Supplier Thermo Scientific Alfa Aesar Supplier No. 036228.36
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

In detinning of iron, metal polishing | CAS: 6009-70-7 | C2H10N2O5 | 142.111 g/mol

In detinning of iron, metal polishing, determination of lead, calcium, and rare earth metalsAmmonium oxalate monohydrate is widely utilized as a buffering reagent and a dispersant to determine the surface interaction of platelets. It finds an application to study

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

  • In detinning of iron, metal polishing, determination of lead, calcium, and rare earth metals
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 6009-70-7
Molecular Formula C2H10N2O5
Molecular Weight (g/mol) 142.111
MDL Number MFCD00149694
InChI Key MSMNVXKYCPHLLN-UHFFFAOYSA-N
Synonym ammonium oxalate monohydrate, ammonium oxalate monohydrate, acs, ethanedioic acid, diammonium salt, monohydrate, diazanium oxalate hydrate, acmc-1aw90, ksc493e5j, diammonium hydrate oxalate, oxalic acid ammonium salt monohydrate
PubChem CID 516808
IUPAC Name diazanium;oxalate;hydrate
SMILES C(=O)(C(=O)[O-])[O-].[NH4+].[NH4+].O

Specifications

Melting Point 70°C (decomposition)
Physical Form Granules
Quantity 500 g
Assay Percent Range 99.0-101.0%
Beilstein 3759830
Merck Index 14,537
Solubility Information Soluble in water; Slightly soluble in alcohol.
Formula Weight 142.11 (124.10anhydrous)
Percent Purity 99.0 to 101.0%
Grade ACS
Density 1.5
Chemical Name or Material Ammonium oxalate, Monohydrate
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.