Learn More
AMPK alpha 2, p Ser345 Antibody (SR2089), Novus Biologicals™
Rabbit Monoclonal Antibody
Brand:  Novus Biologicals NBP3-21563-100ul
 View more versions of this product
Product Code. 18634234
  • € 449.00 / 100µL
Description
AMPK alpha 2 Monoclonal antibody specifically detects AMPK alpha 2 in Human, Mouse, Rat samples. It is validated for Western Blot
TRUSTED_SUSTAINABILITY
Specifications
Specifications
AMPK alpha 2
Monoclonal
Unconjugated
PBS, pH 7.4, 150mM NaCl, 50% glycerol.
Rabbit
Affinity purified
RUO
Primary
Human, Mouse, Rat
Purified
Western Blot
SR2089
Western Blot 1:1000-1:2000
5'-AMP-activated protein kinase catalytic subunit alpha-2, AMPK subunit alpha-2, AMPK2, AMPK5'-AMP-activated protein kinase, catalytic alpha-2 chain, AMPK-alpha-2 chain, EC 2.7.11, EC 2.7.11.1, PRKAA, protein kinase, AMP-activated, alpha 2 catalytic subunit
A synthesized peptide derived from human phospho-AMPK alpha 2 [Ser345] (Uniprot #: P54646)
100 μg
Autophagy, Cancer, Hypoxia, MAP Kinase Signaling, mTOR Pathway, Phospho Specific, Protein Kinase, Signal Transduction, Translation Control
5563
Store at -20 degrees C. Avoid freeze/thaw cycles.