missing translation for 'onlineSavingsMsg'
Learn More
Learn More
AMPK alpha 2, p Ser345 Antibody (SR2089), Novus Biologicals™
Rabbit Monoclonal Antibody
€ 349.00 - € 449.00
Specifications
| Antigen | AMPK alpha 2 |
|---|---|
| Clone | SR2089 |
| Dilution | Western Blot 1:1000-1:2000 |
| Applications | Western Blot |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18634234
|
Novus Biologicals
NBP3-21563-100ul |
100 μg |
€ 449.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18676854
|
Novus Biologicals
NBP3-21563-50ul |
50 μg |
€ 349.00
50µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
AMPK alpha 2 Monoclonal antibody specifically detects AMPK alpha 2 in Human, Mouse, Rat samples. It is validated for Western BlotSpecifications
| AMPK alpha 2 | |
| Western Blot 1:1000-1:2000 | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human, Mouse, Rat | |
| 5'-AMP-activated protein kinase catalytic subunit alpha-2, AMPK subunit alpha-2, AMPK2, AMPK5'-AMP-activated protein kinase, catalytic alpha-2 chain, AMPK-alpha-2 chain, EC 2.7.11, EC 2.7.11.1, PRKAA, protein kinase, AMP-activated, alpha 2 catalytic subunit | |
| A synthesized peptide derived from human phospho-AMPK alpha 2 [Ser345] (Uniprot #: P54646) | |
| Primary | |
| Store at -20 degrees C. Avoid freeze/thaw cycles. |
| SR2089 | |
| Western Blot | |
| Unconjugated | |
| Rabbit | |
| Autophagy, Cancer, Hypoxia, MAP Kinase Signaling, mTOR Pathway, Phospho Specific, Protein Kinase, Signal Transduction, Translation Control | |
| PBS, pH 7.4, 150mM NaCl, 50% glycerol. | |
| 5563 | |
| IgG | |
| Affinity purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title