missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Androgen R/NR3C4 Antibody (AN1-15) - BSA Free, Novus Biologicals™
Rat Monoclonal Antibody
Brand: Novus Biologicals NB300-615-25ug
This item is not returnable.
View return policy
Description
Androgen R/NR3C4 Monoclonal specifically detects Androgen R/NR3C4 in Human, Mouse, Rat, Primate samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunoprecipitation, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen, Gel Super Shift Assays.
Specifications
| Androgen R/NR3C4 | |
| Monoclonal | |
| Unconjugated | |
| P10275 | |
| AR | |
| A fusion protein corresponding to residues 331-572 of human androgen receptor. | |
| 25 μg | |
| Breast Cancer, Cancer, Cellular Markers | |
| 367 | |
| Reconstitute with 50 μL sterilized PBS to a final concentration of 1.0 mg/mL. | |
| Store at -20°C. Avoid freeze/thaw cycles. | |
| IgG |
| Western Blot | |
| AN1-15 | |
| Western Blot 1:50 - 1:500, Immunohistochemistry 2 - 4 μg/mL, Immunoprecipitation 1:10 - 1:500, Immunohistochemistry-Paraffin 2 - 4 μg/mL, Immunohistochemistry-Frozen 2 - 4 μg/mL, Gel Super Shift Assays 1:1 - 1:100, Immunocytochemistry 5 μg/mL | |
| AIS, androgen receptor, DHTRTFM, Dihydrotestosterone receptorHYSP1, HUMARA, NR3C4KD, Nuclear receptor subfamily 3 group C member 4, SMAX1SBMA, spinal and bulbar muscular atrophy | |
| Rat | |
| Protein A purified | |
| RUO | |
| Primary | |
| Androgen receptor. This product does not cross-react with estrogen, progesterone or glucocorticoid receptors. | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction